ENA296275221 Enamine

4-methanesulfonylbenzene-1-carbothioamide

  • MDL number MFCD16622123
  • Cas Number : 875841-36-4
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65334
Availability details for
Quantity
  • Availability
  • Add to Cart