ENA296275222 Enamine

5-(difluoromethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD06195447
  • Cas Number : 866555-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65335
Availability details for
Quantity
  • Availability
  • Add to Cart