ENA296275226 Enamine

2-[(1-methyl-5-phenyl-1H-imidazol-2-yl)sulfanyl]acetic acid

  • MDL number MFCD02595041
  • Cas Number : 462067-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65340
Availability details for
Quantity
  • Availability
  • Add to Cart