ENA296275233 Enamine

3'-chloro-3-methyl-[1,1'-biphenyl]-4-amine

  • MDL number MFCD08701585
  • Cas Number : 893639-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65350
Availability details for
Quantity
  • Availability
  • Add to Cart