ENA296275249 Enamine

1,4-dichlorobut-3-en-2-one

  • MDL number MFCD09038575
  • Cas Number : 15787-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65375

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65375
Availability details for
Quantity
  • Availability
  • Add to Cart