ENA296275250 Enamine

1-methyl-1,2,3,4-tetrahydropyrazine-2,3-dione

  • MDL number MFCD16622127

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65376
Availability details for
Quantity
  • Availability
  • Add to Cart