ENA296275251 Enamine

ethyl 2-[(2-phenylphenyl)amino]acetate

  • MDL number MFCD00779524
  • Cas Number : 39950-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65377
Availability details for
Quantity
  • Availability
  • Add to Cart