ENA296275262 Enamine

2-{4-[2-(pyridin-4-yl)ethyl]piperazin-1-yl}aniline

  • MDL number MFCD16622132
  • Cas Number : 1179164-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65395
Availability details for
Quantity
  • Availability
  • Add to Cart