ENA296275273 Enamine

4-amino-2-chloro-N-(pyridin-3-yl)benzamide

  • MDL number MFCD09047391
  • Cas Number : 19084-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65412
Availability details for
Quantity
  • Availability
  • Add to Cart