ENA296275278 Enamine

3-[(2-phenylethyl)amino]propanoic acid hydrochloride

  • MDL number MFCD11100343
  • Cas Number : 124370-03-2
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65417
Availability details for
Quantity
  • Availability
  • Add to Cart