ENA296275282 Enamine

2-(3-tert-butyl-1,2,4-oxadiazol-5-yl)benzoic acid

  • MDL number MFCD11543487
  • Cas Number : 1038356-45-4
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65421
Availability details for
Quantity
  • Availability
  • Add to Cart