ENA296275284 Enamine

2-chloro-6-(pyrrolidin-1-yl)pyrazine

  • MDL number MFCD09864953
  • Cas Number : 1000339-30-9
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63618
Availability details for
Quantity
  • Availability
  • Add to Cart