ENA296275291 Enamine

5-hydrazinyl-3-phenyl-1,2,4-oxadiazole

  • MDL number MFCD04970007

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65455
Availability details for
Quantity
  • Availability
  • Add to Cart