ENA296275293 Enamine

4-(1H-1,2,3-triazol-1-yl)aniline

  • MDL number MFCD09048597
  • Cas Number : 16279-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65460

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65460
Availability details for
Quantity
  • Availability
  • Add to Cart