ENA296275295 Enamine

2-hydroxy-N-methylacetamide

  • MDL number MFCD02197926
  • Cas Number : 5415-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65463

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65463
Availability details for
Quantity
  • Availability
  • Add to Cart