ENA296275301 Enamine

2-(1-isothiocyanatoethyl)thiophene

  • MDL number MFCD12195531
  • Cas Number : 1153971-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65472
Availability details for
Quantity
  • Availability
  • Add to Cart