ENA296275302 Enamine

3-(2-fluorophenyl)butanoic acid

  • MDL number MFCD11172877
  • Cas Number : 1042815-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65475
Availability details for
Quantity
  • Availability
  • Add to Cart