ENA296275303 Enamine

3-(2,2-difluoroethoxy)-4-methylaniline

  • MDL number MFCD12768442
  • Cas Number : 1183987-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65476
Availability details for
Quantity
  • Availability
  • Add to Cart