ENA296275308 Enamine

methyl 3-(cyclopropylamino)-2-methylpropanoate

  • MDL number MFCD12193193
  • Cas Number : 1154152-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65484
Availability details for
Quantity
  • Availability
  • Add to Cart