ENA296275310 Enamine

2-(dimethylamino)-4-phenyl-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD16622034
  • Cas Number : 50379-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63786
Availability details for
Quantity
  • Availability
  • Add to Cart