ENA296275311 Enamine

(1-amino-2,3-dihydro-1H-inden-1-yl)methanol

  • MDL number MFCD12781979
  • Cas Number : 213924-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65488
Availability details for
Quantity
  • Availability
  • Add to Cart