ENA296275312 Enamine

4-(bromomethyl)-1-phenyl-1H-1,2,3-triazole

  • MDL number MFCD16622139
  • Cas Number : 58432-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65489
Availability details for
Quantity
  • Availability
  • Add to Cart