ENA296275313 Enamine

4-cyclopropyl-5-[(2-fluorophenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD16622140
  • Cas Number : 1240527-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65491
Availability details for
Quantity
  • Availability
  • Add to Cart