ENA296275314 Enamine

5-[(2-chlorophenyl)methyl]-4-cyclopropyl-1,3-thiazol-2-amine

  • MDL number MFCD16622141
  • Cas Number : 1240527-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65494
Availability details for
Quantity
  • Availability
  • Add to Cart