ENA296275316 Enamine

5-methoxy-1,3-dimethyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03791232
  • Cas Number : 26990-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63794

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63794
Availability details for
Quantity
  • Availability
  • Add to Cart