ENA296275317 Enamine

5-(acetamidomethyl)furan-2-carboxylic acid

  • MDL number MFCD13187985
  • Cas Number : 1216078-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65507
Availability details for
Quantity
  • Availability
  • Add to Cart