ENA296275327 Enamine

ethyl 4,6-dimethyl-1H-indole-2-carboxylate

  • MDL number MFCD04966970
  • Cas Number : 95264-40-7
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65520
Availability details for
Quantity
  • Availability
  • Add to Cart