ENA296275329 Enamine

6-(morpholin-4-yl)-9H-purine

  • MDL number MFCD00496816
  • Cas Number : 2846-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63842
Availability details for
Quantity
  • Availability
  • Add to Cart