ENA296275330 Enamine

1-(2,3-dihydro-1H-indol-1-yl)-2-hydroxyethan-1-one

  • MDL number MFCD12138100
  • Cas Number : 1153453-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63855
Availability details for
Quantity
  • Availability
  • Add to Cart