ENA296275332 Enamine

N-(2-aminoethyl)pyrazine-2-carboxamide

  • MDL number MFCD09732292
  • Cas Number : 103879-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63869
Availability details for
Quantity
  • Availability
  • Add to Cart