ENA296275334 Enamine

4-(cyclopropylamino)pyridine-2-carboxylic acid

  • MDL number MFCD11181434
  • Cas Number : 883987-30-2
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65531
Availability details for
Quantity
  • Availability
  • Add to Cart