ENA296275335 Enamine

5-methyl-5-[4-(trifluoromethoxy)phenyl]imidazolidine-2,4-dione

  • MDL number MFCD09933724
  • Cas Number : 1031391-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63939
Availability details for
Quantity
  • Availability
  • Add to Cart