ENA296275341 Enamine

1-[1-(4-chlorophenyl)cyclobutyl]methanamine

  • MDL number MFCD06213128
  • Cas Number : 63010-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65539
Availability details for
Quantity
  • Availability
  • Add to Cart