ENA296275349 Enamine

N-(2-aminophenyl)thiophene-2-carboxamide

  • MDL number MFCD03898669
  • Cas Number : 436095-73-7
  • Molecular Weight : 218.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63982
Availability details for
Quantity
  • Availability
  • Add to Cart