ENA296275350 Enamine

3-(ethylamino)butanoic acid

  • MDL number MFCD12131691
  • Cas Number : 859284-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65556
Availability details for
Quantity
  • Availability
  • Add to Cart