ENA296275351 Enamine

tert-butyl N-(1H-pyrazol-4-ylmethyl)carbamate

  • MDL number MFCD12820298
  • Cas Number : 1107620-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65557

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65557
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart