ENA296275353 Enamine

3-hydroxy-N-(1,3,4-thiadiazol-2-yl)benzamide

  • MDL number MFCD11130526
  • Cas Number : 1019359-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65559
Availability details for
Quantity
  • Availability
  • Add to Cart