ENA296275355 Enamine

1-(1-methyl-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD00540232
  • Cas Number : 19012-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65561
Availability details for
Quantity
  • Availability
  • Add to Cart