ENA296275360 Enamine

(2-methoxyethyl)(1-phenylethyl)amine

  • MDL number MFCD11139348
  • Cas Number : 1019499-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32459
Availability details for
Quantity
  • Availability
  • Add to Cart