ENA296275367 Enamine

(4-methyl-1,3-thiazol-5-yl)methanol

  • MDL number MFCD09032911

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65576

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65576
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart