ENA296275371 Enamine

2-(2,5-dichlorophenoxy)ethan-1-ol

  • MDL number MFCD11614008
  • Cas Number : 101079-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65581
Availability details for
Quantity
  • Availability
  • Add to Cart