ENA296275378 Enamine

6-chloro-N-(3-chlorophenyl)pyridine-3-carboxamide

  • MDL number MFCD07262047
  • Cas Number : 877836-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65589

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65589
Availability details for
Quantity
  • Availability
  • Add to Cart