ENA296275386 Enamine

1-(4-ethylphenyl)-2-methylpropan-1-one

  • MDL number MFCD08457175
  • Cas Number : 68857-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65607
Availability details for
Quantity
  • Availability
  • Add to Cart