ENA296275393 Enamine

methyl 3-(ethylamino)-2-methylpropanoate

  • MDL number MFCD12132809
  • Cas Number : 3440-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65615
Availability details for
Quantity
  • Availability
  • Add to Cart