ENA296275395 Enamine

4-fluoro-3-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD12091825
  • Cas Number : 1156374-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65617
Availability details for
Quantity
  • Availability
  • Add to Cart