ENA296275403 Enamine

3-(2,3-dimethylphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD09941063
  • Cas Number : 1107061-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64231
Availability details for
Quantity
  • Availability
  • Add to Cart