ENA296275404 Enamine

4-bromo-1,2-diethoxybenzene

  • MDL number MFCD09937703
  • Cas Number : 53207-08-2
  • Molecular Weight : 245.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39348
Availability details for
Quantity
  • Availability
  • Add to Cart