ENA296275407 Enamine

(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)methanol

  • MDL number MFCD12137934
  • Cas Number : 99584-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65629

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65629
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart