ENA296275408 Enamine

1-(4-fluorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10696373
  • Cas Number : 214541-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64341
Availability details for
Quantity
  • Availability
  • Add to Cart