ENA296275413 Enamine

2-(pyridin-3-yloxy)acetic acid hydrochloride

  • MDL number MFCD09997611
  • Cas Number : 471894-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64355
Availability details for
Quantity
  • Availability
  • Add to Cart